Assay Details
Target Gene Details
Entrez Gene ID: | 4610 |
Gene Name: | v-myc avian myelocytomatosis viral oncogene lung carcinoma derived homolog |
Gene Aliases: |
L-Myc, LMYC, MYCL1, bHLHe38 |
Location: |
Chr.1:39895424-39902015 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MYCL | NM_001033081.2 | 2 | 1441 | NP_001028253.1 |
| NM_001033082.2 | 3 | 1077 | NP_001028254.2 | |
| AK296078.1 | 3 | 950 | ||
| AK296838.1 | 2 | 624 | ||
| HQ628638.1 | 3 | 760 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2758933 | Chr.1:39759471 - 40194760 on Build GRCh38 | Loss |
|
| esv2666580 | Chr.1:39809257 - 39898016 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map