Assay Details
Target Gene Details
Entrez Gene ID: | 11086 |
Gene Name: | ADAM metallopeptidase domain 29 |
Gene Aliases: |
CT73, svph1 |
Location: |
Chr.4:174918358-174978180 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADAM29 | NM_001130703.1 | 3 | 300 | NP_001124175.1 |
| NM_001130704.1 | 2 | 194 | NP_001124176.1 | |
| NM_001130705.1 | 2 | 194 | NP_001124177.1 | |
| NM_001278125.1 | 4 | 361 | NP_001265054.1 | |
| NM_001278126.1 | 3 | 300 | NP_001265055.1 | |
| NM_001278127.1 | NP_001265056.1 | |||
| NM_014269.4 | 3 | 300 | NP_055084.3 | |
| XM_011531556.1 | 3 | 286 | XP_011529858.1 | |
| XM_011531557.1 | 3 | 286 | XP_011529859.1 | |
| XM_011531559.1 | 3 | 283 | XP_011529861.1 | |
| XM_011531560.1 | 3 | 283 | XP_011529862.1 | |
| XM_011531561.1 | 3 | 283 | XP_011529863.1 | |
| AF134708.1 | 1 | 87 | AAF22163.1 | |
| AK292410.1 | 2 | 147 | ||
| BU176889.1 | 3 | 303 | ||
| DB039551.1 | 2 | 149 | ||
| DB044176.1 | ||||
| DB051105.1 | 3 | 286 | ||
| DB073142.1 | 3 | 301 | ||
| DB078034.1 | 3 | 253 | ||
| DB078125.1 | 3 | 252 | ||
| DB078627.1 | 4 | 314 | ||
| DB080702.1 | 2 | 148 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830156 | Chr.4:174803889 - 174995284 on Build GRCh38 | Gain |
|
| esv3603238 | Chr.4:174843215 - 175044287 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map