Assay Details
Target Gene Details
Entrez Gene ID: | 41 |
Gene Name: | acid sensing ion channel subunit 1 |
Gene Aliases: |
ACCN2, ASIC, BNaC2 |
Location: |
Chr.12:50057637-50083622 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ASIC1 | NM_001095.3 | 6 | 1223 | NP_001086.2 |
| NM_001256830.1 | 4 | 1029 | NP_001243759.1 | |
| NM_020039.3 | 6 | 1223 | NP_064423.2 | |
| NR_046389.1 | 7 | 1305 | ||
| XM_011538350.1 | 7 | 1102 | XP_011536652.1 | |
| XM_011538351.2 | 7 | 1099 | XP_011536653.1 | |
| XM_011538352.1 | 6 | 1009 | XP_011536654.1 | |
| AK290010.1 | 6 | 1008 | ||
| AK293960.1 | 5 | 530 | ||
| AK311047.1 | 4 | 523 | ||
| BC013891.2 | 3 | 237 | AAH13891.2 | |
| BC133707.1 | 5 | 951 | ||
| EU078959.1 | 5 | 927 | ||
| HM991481.1 | 4 | 1029 | ||
| U78180.1 | 6 | 1156 | AAB48980.1 | |
| U78181.1 | 6 | 1156 | AAB48981.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv952179 | Chr.12:50070218 - 50079817 on Build GRCh38 | Deletion |
|
| nsv558840 | Chr.12:50056193 - 50130181 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map