Assay Details
Target Gene Details
Entrez Gene ID: | 124773 |
Gene Name: | chromosome 17 open reading frame 64 |
Gene Aliases: |
- |
Location: |
Chr.17:60421091-60431426 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C17orf64 | NM_181707.2 | NP_859058.2 | ||
| XM_005257033.4 | XP_005257090.1 | |||
| XM_005257034.4 | XP_005257091.1 | |||
| XM_005257035.4 | XP_005257092.1 | |||
| XM_005257036.2 | XP_005257093.1 | |||
| XM_017024166.1 | XP_016879655.1 | |||
| BC036356.1 | AAH36356.1 | |||
| BC048806.1 | AAH48806.1 | |||
| DB448877.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv3273n100 | Chr.17:60293516 - 60519251 on Build GRCh38 | Gain |
|
| nsv519929 | Chr.17:60311418 - 60431257 on Build GRCh38 | Loss |
|
| nsv521420 | Chr.17:60422903 - 60431257 on Build GRCh38 | Loss |
|

Genomic Map