Assay Details
Target Gene Details
Entrez Gene ID: | 22941 |
Gene Name: | SH3 and multiple ankyrin repeat domains 2 |
Gene Aliases: |
AUTS17, CORTBP1, CTTNBP1, ProSAP1, SHANK, SPANK-3 |
Location: |
Chr.11:70467856-71252724 on Build GRCh38 |
Assay Gene Location: | Within Exon 30 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SHANK2 | NM_012309.4 | 25 | 7128 | NP_036441.2 |
| NM_133266.4 | 11 | 5325 | NP_573573.2 | |
| NR_110766.1 | 9 | 2870 | ||
| XM_005277932.3 | 16 | 6133 | XP_005277989.1 | |
| XM_017017387.1 | 26 | 7233 | XP_016872876.1 | |
| XM_017017388.1 | 26 | 7255 | XP_016872877.1 | |
| XM_017017389.1 | 26 | 7401 | XP_016872878.1 | |
| XM_017017390.1 | 13 | 8366 | XP_016872879.1 | |
| AB028945.1 | 8 | 4862 | BAA82974.1 | |
| AB208025.1 | 25 | 7094 | ||
| AB208026.1 | 16 | 5916 | ||
| AB208027.1 | 11 | 5303 | ||
| AK128563.1 | 1 | 2695 | ||
| BX648718.1 | 1 | 3768 |
More Information
Set Membership: |
|

Genomic Map