Assay Details
Target Gene Details
Entrez Gene ID: | 10608 |
Gene Name: | MAX dimerization protein 4 |
Gene Aliases: |
MAD4, MST149, MSTP149, bHLHc12 |
Location: |
Chr.4:2247432-2262143 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MXD4 | NM_006454.2 | 6 | 1976 | NP_006445.1 |
| XM_011513388.2 | 7 | 2481 | XP_011511690.1 | |
| AF082740.1 | 6 | 1976 | AAF98553.1 | |
| AK026392.1 | 1 | 114 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv508990 | Chr.4:2246239 - 2324327 on Build GRCh38 | Insertion |
|
| nsv593377 | Chr.4:2246106 - 2261919 on Build GRCh38 | Loss |
|
| nsv1009347 | Chr.4:2200935 - 2516063 on Build GRCh38 | Gain |
|
| esv2726835 | Chr.4:2019561 - 2959568 on Build GRCh38 | Deletion |
|
| dgv312n21 | Chr.4:2243138 - 2305739 on Build GRCh38 | Loss |
|
| dgv8910n54 | Chr.4:2249015 - 2262496 on Build GRCh38 | Loss |
|
| nsv829838 | Chr.4:2155167 - 2349950 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs74396773] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map