Assay Details
Target Gene Details
Entrez Gene ID: | 26060 |
Gene Name: | adaptor protein, phosphotyrosine interacting with PH domain and leucine zipper 1 |
Gene Aliases: |
APPL, DIP13alpha, MODY14 |
Location: |
Chr.3:57227737-57273471 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| APPL1 | NM_012096.2 | NP_036228.1 | ||
| XM_011533583.2 | XP_011531885.1 | |||
| AB037849.2 | BAA92666.2 | |||
| AF169797.1 | AAF04012.1 | |||
| AF424738.1 | AAL17835.1 | |||
| AK299008.1 | ||||
| AK310716.1 | ||||
| BC028599.2 | AAH28599.1 |
More Information
Set Membership: |
|

Genomic Map