Assay Details
Target Gene Details
Entrez Gene ID: | 9870 |
Gene Name: | apoptosis resistant E3 ubiquitin protein ligase 1 |
Gene Aliases: |
FIEL1, KIAA0317 |
Location: |
Chr.14:74661252-74713118 on Build GRCh38 |
Assay Gene Location: | Within Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AREL1 | NM_001039479.1 | 20 | 4487 | NP_001034568.1 |
| XM_005268244.3 | 19 | 4274 | XP_005268301.1 | |
| XM_006720344.2 | 20 | 4468 | XP_006720407.1 | |
| XM_011537415.2 | 21 | 4560 | XP_011535717.1 | |
| XM_011537416.2 | 19 | 4318 | XP_011535718.1 | |
| XM_011537417.2 | 20 | 4342 | XP_011535719.1 | |
| AB002315.1 | 20 | 4453 | ||
| AK296912.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832831 | Chr.14:74546343 - 74740664 on Build GRCh38 | Loss |
|
| esv3634950 | Chr.14:74661789 - 74676425 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map