Assay Details
Target Gene Details
Entrez Gene ID: | 576 |
Gene Name: | adhesion G protein-coupled receptor B2 |
Gene Aliases: |
BAI2 |
Location: |
Chr.1:31727105-31764063 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADGRB2 | NM_001294335.1 | 11 | 2128 | NP_001281264.1 |
| NM_001294336.1 | 11 | 2128 | NP_001281265.1 | |
| XM_011541848.2 | 10 | 2082 | XP_011540150.1 | |
| XM_011541849.2 | 9 | 1917 | XP_011540151.1 | |
| XM_011541858.2 | 8 | 1081 | XP_011540160.1 | |
| XM_017001899.1 | 10 | 2212 | XP_016857388.1 | |
| XM_017001900.1 | 9 | 2184 | XP_016857389.1 | |
| XM_017001901.1 | 9 | 2184 | XP_016857390.1 | |
| XM_017001902.1 | 9 | 2185 | XP_016857391.1 | |
| XM_017001903.1 | 9 | 2185 | XP_016857392.1 | |
| XM_017001904.1 | 9 | 2181 | XP_016857393.1 | |
| XM_017001905.1 | 9 | 2183 | XP_016857394.1 | |
| XM_017001906.1 | 9 | 2182 | XP_016857395.1 | |
| XM_017001907.1 | 8 | 2016 | XP_016857396.1 | |
| XM_017001908.1 | 8 | 2017 | XP_016857397.1 | |
| XM_017001909.1 | 8 | 2018 | XP_016857398.1 | |
| XM_017001910.1 | 8 | 2016 | XP_016857399.1 | |
| XM_017001911.1 | 8 | 2017 | XP_016857400.1 | |
| XM_017001912.1 | 7 | 1850 | XP_016857401.1 | |
| AB005298.1 | 11 | 2100 | BAA25362.1 | |
| AK294999.1 | ||||
| AK296502.1 | 6 | 760 | ||
| BC136533.1 | 9 | 1796 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv950385 | Chr.1:31655500 - 31800799 on Build GRCh38 | Deletion |
|
| nsv1007786 | Chr.1:31725560 - 31830924 on Build GRCh38 | Gain |
|
| nsv545953 | Chr.1:31699894 - 31749577 on Build GRCh38 | Loss |
|
| esv2758930 | Chr.1:31209533 - 32000730 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map