Assay Details
Target Gene Details
Entrez Gene ID: | 100527960 |
Gene Name: | MROH7-TTC4 readthrough (NMD candidate) |
Gene Aliases: |
HEATR8-TTC4 |
Location: |
Chr.1:54641740-54742657 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 26 - Intron 26 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MROH7-TTC4 | NR_037639.2 | |||
| NR_037640.2 | ||||
| NR_037641.2 | ||||
| AK127008.1 | ||||
| BX648088.1 | ||||
| CR749830.1 | CAH18687.1 |
Target Gene Details
Entrez Gene ID: | 7268 |
Gene Name: | tetratricopeptide repeat domain 4 |
Gene Aliases: |
- |
Location: |
Chr.1:54715822-54742657 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TTC4 | NM_001291333.1 | NP_001278262.1 | ||
| NM_004623.4 | NP_004614.3 | |||
| AF063602.1 | AAG43161.1 | |||
| AF073887.1 | AAD19853.1 | |||
| AK025524.1 | ||||
| AK090964.1 | ||||
| AK222562.1 | BAD96282.1 | |||
| AK292426.1 | ||||
| BC001276.1 | AAH01276.1 | |||
| BT006817.1 | AAP35463.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1004778 | Chr.1:54635180 - 54764639 on Build GRCh38 | Gain |
|
| nsv461561 | Chr.1:54404143 - 54887697 on Build GRCh38 | Gain |
|
| nsv1000673 | Chr.1:54630765 - 54818045 on Build GRCh38 | Gain |
|
| nsv546269 | Chr.1:54593622 - 54757517 on Build GRCh38 | Gain |
|
| dgv38e212 | Chr.1:54627539 - 54817638 on Build GRCh38 | Gain |
|
| nsv817614 | Chr.1:54580862 - 55735351 on Build GRCh38 | Gain |
|
| nsv829904 | Chr.1:54548613 - 54721384 on Build GRCh38 | Loss |
|

Genomic Map