Assay Details
Target Gene Details
Entrez Gene ID: | 22927 |
Gene Name: | hyaluronan binding protein 4 |
Gene Aliases: |
IHABP-4, IHABP4, Ki-1/57, SERBP1L |
Location: |
Chr.9:96450155-96491336 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HABP4 | NM_014282.3 | 1 | 189 | NP_055097.2 |
| AF241831.1 | 1 | 139 | AAF62546.1 | |
| BC018788.1 | 1 | 143 | AAH18788.1 | |
| BI757571.1 | 1 | 212 | ||
| DR006985.1 | 1 | 190 | ||
| HY139664.1 | 1 | 195 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv824998 | Chr.9:96394002 - 96467870 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map