Assay Details
Target Gene Details
Entrez Gene ID: | 124222 |
Gene Name: | progestin and adipoQ receptor family member 4 |
Gene Aliases: |
- |
Location: |
Chr.16:2969245-2973489 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PAQR4 | NM_001284511.1 | 3 | 2034 | NP_001271440.1 |
| NM_001284512.1 | 2 | 1929 | NP_001271441.1 | |
| NM_001284513.1 | 3 | 1657 | NP_001271442.1 | |
| NM_001324118.1 | 4 | 1950 | NP_001311047.1 | |
| NM_152341.4 | 3 | 2151 | NP_689554.2 | |
| AK054564.1 | 3 | 1735 | BAB70758.1 | |
| AK092173.1 | 2 | 1616 | BAC03821.1 | |
| AL832527.1 | 3 | 1764 | ||
| BC033703.1 | 3 | 1809 | AAH33703.1 | |
| BU739514.1 | 1 | 661 |
Target Gene Details
Entrez Gene ID: | 9088 |
Gene Name: | protein kinase, membrane associated tyrosine/threonine 1 |
Gene Aliases: |
MYT1, PPP1R126 |
Location: |
Chr.16:2972791-2980539 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PKMYT1 | NM_001258450.1 | 9 | 1908 | NP_001245379.1 |
| NM_001258451.1 | 9 | 1979 | NP_001245380.1 | |
| NM_004203.4 | 9 | 2115 | NP_004194.3 | |
| NM_182687.2 | 9 | 2153 | NP_872629.1 | |
| XM_011522734.2 | 9 | 2076 | XP_011521036.1 | |
| XM_011522735.2 | 9 | 1902 | XP_011521037.1 | |
| XM_011522736.2 | 8 | 1909 | XP_011521038.1 | |
| AK097642.1 | ||||
| AK098452.1 | ||||
| AK301926.1 | ||||
| AK303005.1 | ||||
| BQ017689.1 | 1 | 73 | ||
| U56816.1 | 8 | 1819 | AAC50949.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv510673 | Chr.16:2919880 - 3043776 on Build GRCh38 | Deletion |
|
| nsv571246 | Chr.16:2942113 - 2973604 on Build GRCh38 | Loss |
|
| nsv827525 | Chr.16:2960856 - 2976333 on Build GRCh38 | Gain |
|
| nsv952906 | Chr.16:2868600 - 3028099 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map