Assay Details
Target Gene Details
Entrez Gene ID: | 57089 |
Gene Name: | ectonucleoside triphosphate diphosphohydrolase 7 |
Gene Aliases: |
LALP1 |
Location: |
Chr.10:99659506-99711245 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ENTPD7 | NM_020354.3 | 3 | 242 | NP_065087.1 |
| AF269255.1 | 3 | 207 | AAF90135.1 | |
| AK055540.1 | 3 | 242 | ||
| AK309735.1 | 4 | 425 | ||
| AK314542.1 | 3 | 300 | ||
| BC119008.1 | 3 | 208 | ||
| BC122857.1 | 3 | 208 |
More Information
Set Membership: |
|

Genomic Map