Assay Details
Target Gene Details
Entrez Gene ID: | 2701 |
Gene Name: | gap junction protein alpha 4 |
Gene Aliases: |
CX37 |
Location: |
Chr.1:34792958-34795747 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GJA4 | NM_002060.2 | 2 | 966 | NP_002051.2 |
| XM_005270750.2 | 2 | 1101 | XP_005270807.1 | |
| XM_017001043.1 | 2 | 1135 | XP_016856532.1 | |
| AF180815.1 | 1 | 878 | AAD56533.1 | |
| AF181620.1 | 1 | 878 | AAD56940.1 | |
| AK291563.1 | 2 | 965 | ||
| AK313477.1 | 2 | 1191 | ||
| BC027889.1 | 2 | 953 | AAH27889.1 | |
| BC072389.1 | 2 | 956 | AAH72389.1 | |
| M96789.2 | 2 | 943 | AAA52558.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1012995 | Chr.1:34786068 - 34884653 on Build GRCh38 | Gain |
|
| nsv945876 | Chr.1:34795129 - 34795810 on Build GRCh38 | Duplication |
|
| esv1003104 | Chr.1:34790757 - 34795666 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map