Assay Details
Target Gene Details
Entrez Gene ID: | 26509 |
Gene Name: | myoferlin |
Gene Aliases: |
FER1L3 |
Location: |
Chr.10:93306429-93482504 on Build GRCh38 |
Assay Gene Location: | Within Exon 55 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MYOF | NM_013451.3 | 51 | 5965 | NP_038479.1 |
| NM_133337.2 | 50 | 5926 | NP_579899.1 | |
| XM_005269693.4 | 52 | 6201 | XP_005269750.1 | |
| XM_005269694.4 | 51 | 6162 | XP_005269751.1 | |
| XM_011539632.2 | 53 | 6395 | XP_011537934.1 | |
| XM_017016068.1 | 51 | 5782 | XP_016871557.1 | |
| XM_017016069.1 | 51 | 5827 | XP_016871558.1 | |
| AB033033.2 | 51 | 5842 | BAA86521.2 | |
| AF182316.1 | 51 | 5938 | AAF27176.1 | |
| AF207990.1 | 50 | 5803 | AAG23737.1 | |
| AK075258.1 | 15 | 1680 | ||
| AK093890.1 | 6 | 1304 | ||
| AL096713.1 | 31 | 3950 | CAB46370.1 | |
| BC052617.1 | 40 | 4478 | AAH52617.1 |
More Information
Set Membership: |
|

Genomic Map