Assay Details
Target Gene Details
Entrez Gene ID: | 10845 |
Gene Name: | caseinolytic mitochondrial matrix peptidase chaperone subunit |
Gene Aliases: |
- |
Location: |
Chr.15:65148219-65185420 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CLPX | NM_006660.4 | NP_006651.2 | ||
| NR_133680.1 | ||||
| XM_011521164.2 | XP_011519466.1 | |||
| AJ006267.3 | CAA06933.2 | |||
| AK292316.1 | ||||
| AK308133.1 | ||||
| AL136922.1 | CAB66856.1 | |||
| BC130373.1 | ||||
| BC136487.1 |
More Information
Set Membership: |
|

Genomic Map