Assay Details
Target Gene Details
Entrez Gene ID: | 55333 |
Gene Name: | synaptojanin 2 binding protein |
Gene Aliases: |
ARIP2, OMP25 |
Location: |
Chr.14:70366496-70417090 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SYNJ2BP | NM_018373.2 | 4 | 706 | NP_060843.2 |
| AK002133.1 | 4 | 661 | BAA92098.1 | |
| AK311719.1 | 3 | 540 | ||
| AY534623.1 | 4 | 675 | AAT01566.1 | |
| BC007704.2 | AAH07704.1 |
Target Gene Details
Entrez Gene ID: | 100529257 |
Gene Name: | SYNJ2BP-COX16 readthrough |
Gene Aliases: |
- |
Location: |
Chr.14:70325081-70417090 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SYNJ2BP-COX16 | NM_001202547.2 | NP_001189476.1 | ||
| NM_001202548.2 | NP_001189477.1 | |||
| NM_001202549.2 | NP_001189478.1 | |||
| FY210787.1 | ||||
| FY211163.1 | ||||
| FY213292.1 | ||||
| FY213836.1 |
More Information
Set Membership: |
|

Genomic Map