Assay Details
Target Gene Details
Entrez Gene ID: | 25960 |
Gene Name: | adhesion G protein-coupled receptor A2 |
Gene Aliases: |
GPR124, TEM5 |
Location: |
Chr.8:37796883-37843986 on Build GRCh38 |
Assay Gene Location: | Within Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADGRA2 | NM_032777.9 | 19 | 5912 | NP_116166.9 |
| XM_011544481.2 | 19 | 5981 | XP_011542783.1 | |
| XM_011544482.2 | 18 | 5909 | XP_011542784.1 | |
| XM_011544483.2 | 18 | 5822 | XP_011542785.1 | |
| AB040964.2 | 16 | 5238 | BAA96055.2 | |
| AF378755.1 | 19 | 5539 | AAL11992.1 | |
| AI346793.1 | 1 | 139 | ||
| AK027296.1 | 6 | 1952 | BAB55022.1 | |
| AK074099.2 | 19 | 5443 | ||
| AL110244.1 | 1 | 2533 | CAB53694.1 | |
| AL137610.1 | 1 | 1011 | ||
| AL832524.1 | 3 | 2979 | CAD38629.1 | |
| AL832959.1 | 12 | 4582 | CAH56283.1 | |
| BC146774.1 | 16 | 5238 |
Target Gene Details
Entrez Gene ID: | 55290 |
Gene Name: | BRF2, RNA polymerase III transcription initiation factor 50 kDa subunit |
Gene Aliases: |
BRFU, TFIIIB50 |
Location: |
Chr.8:37843880-37849913 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BRF2 | NM_018310.3 | NP_060780.2 | ||
| AA496239.1 | ||||
| AF130058.1 | AAG35486.1 | |||
| AF206673.2 | AAG35669.2 | |||
| AK001914.1 | BAA91975.1 | |||
| BC010648.1 | AAH10648.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv509261 | Chr.8:37767151 - 37881434 on Build GRCh38 | Insertion |
|
| nsv528598 | Chr.8:37803553 - 37859603 on Build GRCh38 | Loss |
|
| nsv951126 | Chr.8:37827983 - 37845682 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map