Assay Details
Target Gene Details
Entrez Gene ID: | 8448 |
Gene Name: | double C2 domain alpha |
Gene Aliases: |
Doc2 |
Location: |
Chr.16:30005514-30023280 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DOC2A | NM_001282062.1 | NP_001268991.1 | ||
| NM_001282063.1 | NP_001268992.1 | |||
| NM_001282068.1 | NP_001268997.1 | |||
| NM_003586.2 | NP_003577.2 | |||
| NR_104089.1 | ||||
| NR_104090.1 | ||||
| XM_011545975.1 | XP_011544277.1 | |||
| XM_017023776.1 | XP_016879265.1 | |||
| XM_017023777.1 | XP_016879266.1 | |||
| XM_017023778.1 | XP_016879267.1 | |||
| AK090829.1 | ||||
| BC055284.1 | AAH55284.1 |
Target Gene Details
Entrez Gene ID: | 283899 |
Gene Name: | INO80 complex subunit E |
Gene Aliases: |
CCDC95 |
Location: |
Chr.16:29996209-30005794 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| INO80E | NM_001304562.1 | 6 | 840 | NP_001291491.1 |
| NM_001304563.1 | 8 | 1047 | NP_001291492.1 | |
| NM_173618.2 | 7 | 957 | NP_775889.1 | |
| NR_130786.1 | 8 | 1249 | ||
| NR_130787.1 | 7 | 1159 | ||
| NR_134855.1 | 11 | 1737 | ||
| XM_011545809.2 | 7 | 794 | XP_011544111.1 | |
| AI686175.1 | 1 | 277 | ||
| AK074033.1 | 1 | 4149 | BAB84859.1 | |
| AK074137.1 | 7 | 4257 | BAB84963.1 | |
| AK075133.1 | 7 | 956 | BAC11424.1 | |
| AK097889.1 | 1 | 1696 | ||
| AK130409.1 | 7 | 948 | ||
| AY189289.1 | 10 | 2835 | AAO86733.1 | |
| BC047712.2 | 7 | 918 | AAH47712.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3638378 | Chr.16:29973809 - 30006402 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map