Assay Details
Target Gene Details
Entrez Gene ID: | 11095 |
Gene Name: | ADAM metallopeptidase with thrombospondin type 1 motif 8 |
Gene Aliases: |
ADAM-TS8, METH2 |
Location: |
Chr.11:130404923-130428993 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADAMTS8 | NM_007037.5 | 9 | 2940 | NP_008968.4 |
| XM_017017145.1 | 9 | 2412 | XP_016872634.1 | |
| AB209717.1 | 5 | 821 | BAD92954.1 | |
| AF060153.1 | 9 | 2941 | AAD48081.1 | |
| AF175283.1 | AAF25806.1 | |||
| AK123706.1 | 4 | 1180 | ||
| BC089435.1 | 10 | 2320 | AAH89435.1 | |
| BC113874.1 | 10 | 1595 | AAI13875.1 | |
| BC114624.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759871 | Chr.11:130099327 - 130472822 on Build GRCh38 | Loss |
|
| nsv508660 | Chr.11:130397628 - 130503590 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map