Assay Details
Target Gene Details
Entrez Gene ID: | 51268 |
Gene Name: | pipecolic acid and sarcosine oxidase |
Gene Aliases: |
LPIPOX |
Location: |
Chr.17:29042900-29057227 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PIPOX | NM_016518.2 | 8 | 1564 | NP_057602.2 |
| XM_017024722.1 | 7 | 1163 | XP_016880211.1 | |
| AF134593.1 | 8 | 1281 | AAF37331.1 | |
| AF136970.1 | 8 | 1250 | AAG49431.1 | |
| AK022773.1 | 8 | 1329 | ||
| AK027498.1 | 8 | 1288 | ||
| BC008960.2 | 8 | 1564 | AAH08960.1 | |
| BC027622.2 | 8 | 1300 | AAH27622.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833408 | Chr.17:28913470 - 29105450 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map