Assay Details
Target Gene Details
Entrez Gene ID: | 113402 |
Gene Name: | SFT2 domain containing 1 |
Gene Aliases: |
C6orf83, pRGR1 |
Location: |
Chr.6:166319728-166342606 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SFT2D1 | NM_145169.2 | NP_660152.1 | ||
| NR_130112.1 | ||||
| NR_130113.1 | ||||
| NR_130114.1 | ||||
| AF041429.1 | AAF18564.1 | |||
| AK021489.1 | ||||
| AK090795.1 | 1 | 1511 | ||
| AL833217.1 | ||||
| AW964442.1 | ||||
| BC018969.1 | AAH18969.1 | |||
| BC028388.1 | AAH28388.1 | |||
| BF223532.1 | ||||
| BG706927.1 | ||||
| BI597156.1 |
More Information
Set Membership: |
|

Genomic Map