Assay Details
Target Gene Details
Entrez Gene ID: | 6368 |
Gene Name: | C-C motif chemokine ligand 23 |
Gene Aliases: |
CK-BETA-8, CKb8, Ckb-8, Ckb-8-1, MIP-3, MIP3, MPIF-1, SCYA23, hmrp-2a |
Location: |
Chr.17:36013056-36018179 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCL23 | NM_005064.5 | NP_005055.3 | ||
| NM_145898.3 | NP_665905.2 | |||
| BC093970.1 | AAH93970.1 | |||
| BC143310.1 | ||||
| U58913.1 | AAD10846.1 | |||
| U67128.1 | AAD00161.1 | |||
| U85767.1 | AAB51134.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1059795 | Chr.17:35828724 - 36157706 on Build GRCh38 | Gain |
|
| nsv833427 | Chr.17:35958281 - 36115227 on Build GRCh38 | Loss |
|
| esv3640462 | Chr.17:36000619 - 36055115 on Build GRCh38 | Gain |
|
| nsv1055760 | Chr.17:36001422 - 36062566 on Build GRCh38 | Gain |
|
| nsv833424 | Chr.17:35911883 - 36092128 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map