Assay Details
Target Gene Details
Entrez Gene ID: | 1891 |
Gene Name: | enoyl-CoA hydratase 1 |
Gene Aliases: |
HPXEL |
Location: |
Chr.19:38815422-38831857 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ECH1 | NM_001398.2 | NP_001389.2 | ||
| XM_017026448.1 | XP_016881937.1 | |||
| AK291860.1 | ||||
| AK301211.1 | ||||
| BC011792.2 | AAH11792.1 | |||
| BC017408.1 | AAH17408.1 | |||
| BG527600.1 | ||||
| U16660.1 | AAC50222.1 |
More Information
Set Membership: |
|

Genomic Map