Assay Details
Target Gene Details
Entrez Gene ID: | 100885848 |
Gene Name: | prostaglandin E synthase 3 like |
Gene Aliases: |
- |
Location: |
Chr.17:42968088-42980528 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PTGES3L | NM_001142653.1 | NP_001136125.1 | ||
| NM_001142654.1 | NP_001136126.1 | |||
| NM_001261430.1 | NP_001248359.1 | |||
| AK056965.1 | ||||
| AK093402.1 | 1 | 670 | ||
| BG776211.1 | ||||
| DA900052.1 | ||||
| DC329857.1 |
Target Gene Details
Entrez Gene ID: | 100885850 |
Gene Name: | PTGES3L-AARSD1 readthrough |
Gene Aliases: |
- |
Location: |
Chr.17:42950526-42980528 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PTGES3L-AARSD1 | NM_001136042.2 | NP_001129514.2 | ||
| NM_025267.3 | NP_079543.1 | |||
| AF039235.1 | ||||
| AK094084.1 | ||||
| AK295447.1 | ||||
| BC004172.2 | AAH04172.1 | |||
| BC019324.1 | AAH19324.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv2057 | Chr.17:42946856 - 42980330 on Build GRCh38 | Insertion |
|
| esv2672913 | Chr.17:42964983 - 43322347 on Build GRCh38 | Deletion |
|
| nsv483009 | Chr.17:42829470 - 43006887 on Build GRCh38 | Gain |
|
| nsv575052 | Chr.17:42907778 - 43019464 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map