Assay Details
Target Gene Details
Entrez Gene ID: | 3003 |
Gene Name: | granzyme K |
Gene Aliases: |
TRYP2 |
Location: |
Chr.5:55024279-55034132 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GZMK | NM_002104.2 | NP_002095.1 | ||
| AK312074.1 | ||||
| BC035802.1 | AAH35802.1 | |||
| U26174.1 | AAA74578.1 | |||
| U35237.1 | AAA79063.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830301 | Chr.5:54930046 - 55127677 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map