Assay Details
Target Gene Details
Entrez Gene ID: | 79183 |
Gene Name: | alpha tocopherol transfer protein like |
Gene Aliases: |
C20orf121 |
Location: |
Chr.20:44475886-44494603 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TTPAL | NM_001039199.2 | 2 | 365 | NP_001034288.1 |
| NM_001261839.1 | 2 | 365 | NP_001248768.1 | |
| NM_024331.4 | 3 | 389 | NP_077307.2 | |
| XM_011529043.2 | 3 | 501 | XP_011527345.1 | |
| XM_011529044.2 | 3 | 1116 | XP_011527346.1 | |
| XM_011529045.2 | 2 | 365 | XP_011527347.1 | |
| XM_017028063.1 | 3 | 380 | XP_016883552.1 | |
| AK307236.1 | 1 | 275 | ||
| AK314045.1 | 2 | 324 | ||
| BC003071.2 | 3 | 368 | ||
| BP351602.1 | 2 | 367 | ||
| BQ646722.1 | 2 | 331 | ||
| CR936621.1 | 2 | 319 | CAI56764.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv586065 | Chr.20:44261867 - 44505244 on Build GRCh38 | Gain |
|
| nsv1061719 | Chr.20:44415722 - 44602319 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map