Assay Details
Target Gene Details
Entrez Gene ID: | 148304 |
Gene Name: | chromosome 1 open reading frame 74 |
Gene Aliases: |
URLC4 |
Location: |
Chr.1:209779208-209784730 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C1orf74 | NM_152485.3 | 2 | 1654 | NP_689698.1 |
| AB101206.1 | 2 | 1654 | ||
| AK057807.1 | 2 | 1483 | BAB71584.1 | |
| BC039719.1 | 2 | 1469 | AAH39719.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv4343 | Chr.1:209756587 - 209801532 on Build GRCh38 | Deletion |
|
| nsv1006632 | Chr.1:209712857 - 209797010 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs144888845] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map