Assay Details
Target Gene Details
Entrez Gene ID: | 4209 |
Gene Name: | myocyte enhancer factor 2D |
Gene Aliases: |
- |
Location: |
Chr.1:156463721-156500842 on Build GRCh38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MEF2D | NM_001271629.1 | 11 | 3555 | NP_001258558.1 |
| NM_005920.3 | 12 | 3857 | NP_005911.1 | |
| XM_005245169.4 | 12 | 3839 | XP_005245226.1 | |
| XM_005245170.3 | 12 | 3864 | XP_005245227.1 | |
| XM_006711330.3 | 12 | 3568 | XP_006711393.1 | |
| XM_006711332.3 | 12 | 3852 | XP_006711395.1 | |
| XM_006711333.2 | 11 | 3831 | XP_006711396.1 | |
| XM_006711334.3 | 11 | 3829 | XP_006711397.1 | |
| XM_011509569.2 | 12 | 3889 | XP_011507871.1 | |
| XM_017001314.1 | 11 | 3973 | XP_016856803.1 | |
| XM_017001315.1 | 11 | 3568 | XP_016856804.1 | |
| AB210005.1 | 11 | 4107 | BAE06087.1 | |
| BC054520.1 | 12 | 3752 | AAH54520.1 | |
| BX640792.1 | 1 | 298 | ||
| BX647226.1 | 1 | 1594 |
More Information
Set Membership: |
|

Genomic Map