Assay Details
Target Gene Details
Entrez Gene ID: | 92106 |
Gene Name: | oxidoreductase NAD binding domain containing 1 |
Gene Aliases: |
- |
Location: |
Chr.3:16265160-16386979 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OXNAD1 | NM_138381.3 | 3 | 579 | NP_612390.1 |
| XM_005265559.3 | 3 | 560 | XP_005265616.1 | |
| XM_006713405.1 | 3 | 543 | XP_006713468.1 | |
| XM_006713406.2 | 3 | 528 | XP_006713469.1 | |
| XM_011534230.2 | 3 | 560 | XP_011532532.1 | |
| XM_011534232.2 | 3 | 560 | XP_011532534.1 | |
| XM_011534233.2 | 3 | 560 | XP_011532535.1 | |
| XM_011534234.2 | 3 | 560 | XP_011532536.1 | |
| XM_011534235.2 | 3 | 560 | XP_011532537.1 | |
| XM_017007485.1 | 3 | 543 | XP_016862974.1 | |
| XM_017007486.1 | 3 | 575 | XP_016862975.1 | |
| XM_017007487.1 | 3 | 575 | XP_016862976.1 | |
| XM_017007488.1 | 3 | 575 | XP_016862977.1 | |
| XM_017007489.1 | 3 | 575 | XP_016862978.1 | |
| XM_017007490.1 | 3 | 575 | XP_016862979.1 | |
| XM_017007491.1 | 3 | 575 | XP_016862980.1 | |
| XM_017007492.1 | 3 | 314 | XP_016862981.1 | |
| XM_017007493.1 | 3 | 528 | XP_016862982.1 | |
| XM_017007494.1 | 3 | 528 | XP_016862983.1 | |
| XM_017007495.1 | 3 | 525 | XP_016862984.1 | |
| XM_017007496.1 | 3 | 525 | XP_016862985.1 | |
| AB209556.1 | 1 | 73 | BAD92793.1 | |
| AK298829.1 | 2 | 134 | ||
| AK307258.1 | 3 | 506 | ||
| AL583453.3 | 3 | 538 | ||
| BC008322.1 | 3 | 506 | AAH08322.1 | |
| DQ364598.1 | 3 | 537 | ABC95193.1 |
More Information
Set Membership: |
|

Genomic Map