Assay Details
Target Gene Details
Entrez Gene ID: | 23767 |
Gene Name: | fibronectin leucine rich transmembrane protein 3 |
Gene Aliases: |
HH21 |
Location: |
Chr.20:14322985-14337847 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FLRT3 | NM_013281.3 | 2 | 406 | NP_037413.1 |
| NM_198391.2 | 3 | 600 | NP_938205.1 | |
| XM_005260682.4 | 3 | 1118 | XP_005260739.1 | |
| XM_011529204.2 | 4 | 1255 | XP_011527506.1 | |
| XM_011529205.2 | 4 | 917 | XP_011527507.1 | |
| AB040902.1 | 2 | 294 | ||
| AF169677.1 | 2 | 303 | AAF28461.1 | |
| AK027297.1 | 2 | 317 | BAB55023.1 | |
| AK027694.1 | BAB55303.1 | |||
| AK074883.1 | 2 | 347 | BAC11265.1 | |
| AK074909.1 | 2 | 275 | BAC11284.1 | |
| AL529481.3 | 3 | 557 | ||
| AY358319.1 | 1 | 140 | AAQ88685.1 | |
| BC020870.1 | 2 | 313 | AAH20870.1 | |
| CD654700.1 | 2 | 367 | ||
| DA502792.1 | 2 | 406 |
Target Gene Details
Entrez Gene ID: | 140733 |
Gene Name: | MACRO domain containing 2 |
Gene Aliases: |
C20orf133 |
Location: |
Chr.20:13995476-16053197 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MACROD2 | NM_080676.5 | NP_542407.2 | ||
| XM_017027677.1 | XP_016883166.1 | |||
| AK131348.1 | BAD18504.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3893290 | Chr.20:14153723 - 14357847 on Build GRCh38 | Gain |
|
| nsv585439 | Chr.20:14209133 - 14336744 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map