Assay Details
Target Gene Details
Entrez Gene ID: | 761 |
Gene Name: | carbonic anhydrase 3 |
Gene Aliases: |
CAIII, Car3 |
Location: |
Chr.8:85438827-85449040 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CA3 | NM_005181.3 | NP_005172.1 | ||
| AK096880.1 | ||||
| AK300670.1 | ||||
| AK300677.1 | ||||
| AK313254.1 | ||||
| BC004897.2 | AAH04897.1 | |||
| DA421884.1 | ||||
| EU794596.1 |
Target Gene Details
Entrez Gene ID: | 100996348 |
Gene Name: | CA3 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.8:85441806-85464915 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CA3-AS1 | NR_121630.1 | |||
| NR_121631.1 | ||||
| NR_121632.1 | ||||
| BM713112.1 | ||||
| BM717037.1 | ||||
| DA940436.1 | ||||
| DB177224.1 | ||||
| DB196812.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831377 | Chr.8:85350878 - 85531912 on Build GRCh38 | Gain |
|
| nsv1028148 | Chr.8:85398341 - 85640901 on Build GRCh38 | Gain |
|
| esv2761206 | Chr.8:85398353 - 85556639 on Build GRCh38 | Gain |
|
| nsv483126 | Chr.8:83687766 - 85887771 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map