Assay Details
Target Gene Details
Entrez Gene ID: | 84873 |
Gene Name: | adhesion G protein-coupled receptor G7 |
Gene Aliases: |
GPR128 |
Location: |
Chr.3:100609589-100696174 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 6 - Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADGRG7 | NM_001308362.1 | NP_001295291.1 | ||
| NM_032787.2 | NP_116176.2 | |||
| XM_011513245.2 | XP_011511547.1 | |||
| AK027360.1 | BAB55061.1 | |||
| AK172769.1 | BAD18752.1 | |||
| AK295345.1 | ||||
| AY181246.1 | AAO27358.1 | |||
| BC113451.1 | ||||
| BC130388.1 | ||||
| DA640995.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2751970 | Chr.3:100623466 - 100723634 on Build GRCh38 | Gain |
|
| dgv613n27 | Chr.3:100591968 - 100714748 on Build GRCh38 | Gain |
|
| dgv8477n54 | Chr.3:100626357 - 100699750 on Build GRCh38 | Gain |
|
| dgv182e55 | Chr.3:100580466 - 100760540 on Build GRCh38 | Gain |
|
| nsv1010619 | Chr.3:100634965 - 100670413 on Build GRCh38 | Gain |
|
| dgv4831n100 | Chr.3:100621212 - 100709093 on Build GRCh38 | Gain |
|
| esv2763708 | Chr.3:100599084 - 100723665 on Build GRCh38 | Gain |
|
| nsv516418 | Chr.3:100605598 - 100714748 on Build GRCh38 | Gain |
|
| dgv4830n100 | Chr.3:100605431 - 100729339 on Build GRCh38 | Gain |
|
| dgv183e55 | Chr.3:100623649 - 100774218 on Build GRCh38 | Gain |
|
| dgv1397e212 | Chr.3:100607666 - 100723653 on Build GRCh38 | Gain |
|
| nsv470794 | Chr.3:100631901 - 100714748 on Build GRCh38 | Gain |
|
| dgv917e214 | Chr.3:100604090 - 100730204 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map