Assay Details
Target Gene Details
Entrez Gene ID: | 124944 |
Gene Name: | chromosome 17 open reading frame 49 |
Gene Aliases: |
BAP18, HEPIS |
Location: |
Chr.17:7014737-7017524 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C17orf49 | NM_001142798.2 | NP_001136270.1 | ||
| NM_001142799.2 | NP_001136271.1 | |||
| NM_174893.3 | NP_777553.1 | |||
| AK055800.1 | 2 | 1579 | ||
| AK295795.1 | ||||
| BC040036.1 | AAH40036.1 | |||
| BM011108.1 | ||||
| BU596784.1 | ||||
| BU730728.1 | ||||
| CA449884.1 | ||||
| CB119197.1 | ||||
| CN280697.1 |
Target Gene Details
Entrez Gene ID: | 100506755 |
Gene Name: | mir-497-195 cluster host gene |
Gene Aliases: |
MIR195HG |
Location: |
Chr.17:7015818-7019654 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MIR497HG | NR_038310.1 |
Target Gene Details
Entrez Gene ID: | 100529209 |
Gene Name: | RNASEK-C17orf49 readthrough |
Gene Aliases: |
- |
Location: |
Chr.17:7012417-7017524 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RNASEK-C17orf49 | NR_037717.1 | |||
| BC040148.1 | AAH40148.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2422288 | Chr.17:6789933 - 7646709 on Build GRCh38 | Deletion |
|
| nsv952117 | Chr.17:7014482 - 7028381 on Build GRCh38 | Deletion |
|
| nsv457659 | Chr.17:7010333 - 7689462 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map