Assay Details
Target Gene Details
Entrez Gene ID: | 23404 |
Gene Name: | exosome component 2 |
Gene Aliases: |
RRP4, Rrp4p, hRrp4p, p7 |
Location: |
Chr.9:130693760-130704894 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EXOSC2 | NM_001282708.1 | 8 | 1269 | NP_001269637.1 |
| NM_001282709.1 | 8 | 1257 | NP_001269638.1 | |
| NM_014285.6 | 9 | 1347 | NP_055100.2 | |
| NR_104230.1 | 8 | 1853 | ||
| XM_005272176.2 | 9 | 1345 | XP_005272233.1 | |
| XM_006717023.2 | 8 | 1255 | XP_006717086.1 | |
| XM_017014558.1 | 8 | 1879 | XP_016870047.1 | |
| AK001916.1 | 9 | 1336 | BAA91977.1 | |
| AK022460.1 | 9 | 1333 | BAB14043.1 | |
| AK090512.1 | 8 | 1879 | ||
| AK092727.1 | 3 | 2113 | ||
| BC000747.2 | 9 | 1317 | AAH00747.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3621866 | Chr.9:130695968 - 131205700 on Build GRCh38 | Gain |
|
| nsv831735 | Chr.9:130598942 - 130791581 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map