Assay Details
Target Gene Details
Entrez Gene ID: | 221188 |
Gene Name: | adhesion G protein-coupled receptor G5 |
Gene Aliases: |
GPR114, PGR27 |
Location: |
Chr.16:57529339-57578182 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADGRG5 | NM_001304376.2 | 4 | 360 | NP_001291305.1 |
| NM_001318481.1 | 4 | 360 | NP_001305410.1 | |
| NM_153837.3 | 4 | 832 | NP_722579.1 | |
| XM_011522949.2 | 4 | 376 | XP_011521251.1 | |
| XM_011522950.2 | 4 | 402 | XP_011521252.1 | |
| AK128298.1 | 4 | 511 | ||
| AK160368.1 | 4 | 342 | BAD18711.1 | |
| AK292783.1 | 4 | 352 | ||
| AK301357.1 | 4 | 338 | ||
| AK303661.1 | 4 | 352 | ||
| AY140956.1 | 3 | 257 | AAN46670.1 | |
| AY358447.1 | 4 | 339 | AAQ88812.1 | |
| BC032401.1 | 4 | 780 | AAH32401.1 | |
| BM193863.1 | 4 | 354 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833245 | Chr.16:57434073 - 57584619 on Build GRCh38 | Loss |
|
| nsv952036 | Chr.16:57561989 - 57582388 on Build GRCh38 | Deletion |
|
| nsv1055537 | Chr.16:57531862 - 57570612 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map