Assay Details
Target Gene Details
Entrez Gene ID: | 1508 |
Gene Name: | cathepsin B |
Gene Aliases: |
APPS, CPSB |
Location: |
Chr.8:11842524-11868150 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 13 - Exon 13 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1034132 | Chr.8:11799524 - 12340733 on Build GRCh38 | Gain |
|
| nsv610293 | Chr.8:11841238 - 12072225 on Build GRCh38 | Gain |
|
| esv34042 | Chr.8:11840828 - 11869308 on Build GRCh38 | Loss |
|
| nsv1018042 | Chr.8:11809987 - 12024876 on Build GRCh38 | Gain |
|
| esv3572516 | Chr.8:11844191 - 11853761 on Build GRCh38 | Loss |
|
| nsv1032739 | Chr.8:11786294 - 11858964 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map