Assay Details
Target Gene Details
Entrez Gene ID: | 51611 |
Gene Name: | diphthamide biosynthesis 5 |
Gene Aliases: |
AD-018, CGI-30, HSPC143, NPD015 |
Location: |
Chr.1:100989623-101025806 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DPH5 | NM_001077394.1 | 8 | 867 | NP_001070862.1 |
| NM_001077395.1 | 8 | 846 | NP_001070863.1 | |
| NM_015958.2 | 8 | 849 | NP_057042.2 | |
| XM_005270938.1 | 7 | 745 | XP_005270995.1 | |
| XM_005270939.4 | 7 | 657 | XP_005270996.1 | |
| XM_005270941.3 | 7 | 632 | XP_005270998.1 | |
| XM_006710683.3 | 6 | 504 | XP_006710746.1 | |
| XM_011541569.1 | 7 | 742 | XP_011539871.1 | |
| XM_011541570.2 | 7 | 649 | XP_011539872.1 | |
| XM_017001460.1 | 8 | 869 | XP_016856949.1 | |
| XM_017001461.1 | 7 | 656 | XP_016856950.1 | |
| XM_017001462.1 | 7 | 651 | XP_016856951.1 | |
| AF132964.1 | 8 | 854 | AAD27739.1 | |
| AF157319.1 | 7 | 664 | AAF67485.1 | |
| AF161492.1 | 7 | 705 | AAF29107.1 | |
| AF248965.1 | 8 | 818 | AAG44563.1 | |
| AK125485.1 | 9 | 1218 | ||
| AK225469.1 | 8 | 824 | ||
| AK225707.1 | 8 | 824 | ||
| AK225951.1 | 8 | 824 | ||
| AK289351.1 | 8 | 835 | ||
| AK298181.1 | 6 | 571 | ||
| BC009620.2 | 2 | 109 | AAH09620.2 | |
| BC053857.1 | 8 | 815 | AAH53857.1 | |
| BI907367.1 |
Target Gene Details
Entrez Gene ID: | 148867 |
Gene Name: | solute carrier family 30 member 7 |
Gene Aliases: |
ZNT7, ZnT-7, ZnTL2 |
Location: |
Chr.1:100896076-100996913 on Build GRCh38 |
Assay Gene Location: | Within Intron 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC30A7 | XM_017000400.1 | XP_016855889.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv522064 | Chr.1:100715786 - 101074310 on Build GRCh38 | Gain |
|
| esv3586958 | Chr.1:100987009 - 100992949 on Build GRCh38 | Loss |
|
| esv3586960 | Chr.1:100988657 - 100992115 on Build GRCh38 | Gain |
|
| esv2668878 | Chr.1:100986901 - 100992861 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map