Assay Details
Target Gene Details
Entrez Gene ID: | 9476 |
Gene Name: | napsin A aspartic peptidase |
Gene Aliases: |
KAP, Kdap, NAP1, NAPA, SNAPA |
Location: |
Chr.19:50358472-50369331 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NAPSA | NM_004851.2 | NP_004842.1 | ||
| XM_011527540.1 | XP_011525842.1 | |||
| XM_017027512.1 | XP_016883001.1 | |||
| AF090386.1 | AAD04917.1 | |||
| AF098484.1 | AAD13215.1 | |||
| AF200345.1 | AAF17081.1 | |||
| AK055569.1 | 1 | 1070 | ||
| AK308013.1 | ||||
| AK314364.1 | ||||
| BC017842.1 | AAH17842.1 | |||
| BI770845.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1066389 | Chr.19:50319151 - 50453251 on Build GRCh38 | Gain |
|
| nsv2529 | Chr.19:50336652 - 50368507 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map