Assay Details
Target Gene Details
Entrez Gene ID: | 10745 |
Gene Name: | putative homeodomain transcription factor 1 |
Gene Aliases: |
PHTF |
Location: |
Chr.1:113697202-113759888 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 13 - Exon 14 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3587245 | Chr.1:113699035 - 113723249 on Build GRCh38 | Loss |
|
| esv3587246 | Chr.1:113705344 - 113706268 on Build GRCh38 | Loss |
|
| esv3587240 | Chr.1:113533246 - 114158910 on Build GRCh38 | Loss |
|
| esv3587239 | Chr.1:113510852 - 113866662 on Build GRCh38 | Loss |
|
| esv23869 | Chr.1:113320330 - 114358495 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map