Assay Details
Target Gene Details
Entrez Gene ID: | 11282 |
Gene Name: | mannosyl (alpha-1,3-)-glycoprotein beta-1,4-N-acetylglucosaminyltransferase, isozyme B |
Gene Aliases: |
GNT-IV, GNT-IVB |
Location: |
Chr.5:179797597-179806952 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MGAT4B | NM_014275.4 | NP_055090.1 | ||
| NM_054013.3 | NP_463459.1 | |||
| XM_017008998.1 | 14 | 1779 | XP_016864487.1 | |
| AB000624.1 | BAA83464.1 | |||
| AK023137.1 | ||||
| AK225490.1 | ||||
| AK225492.1 | ||||
| AL162067.1 | CAB82404.2 | |||
| AY358984.1 | AAQ89343.1 | |||
| BC009464.1 | AAH09464.2 | |||
| BC051835.1 | AAH51835.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv600659 | Chr.5:179787250 - 179798594 on Build GRCh38 | Loss |
|
| nsv518964 | Chr.5:179766597 - 179800286 on Build GRCh38 | Loss |
|
| nsv469579 | Chr.5:179757830 - 179933191 on Build GRCh38 | Loss |
|
| dgv10241n54 | Chr.5:179794788 - 179802039 on Build GRCh38 | Loss |
|
| nsv823361 | Chr.5:179786323 - 179824243 on Build GRCh38 | Loss |
|
| nsv600661 | Chr.5:179793009 - 179801623 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map