Assay Details
Target Gene Details
Entrez Gene ID: | 28971 |
Gene Name: | adipogenesis associated Mth938 domain containing |
Gene Aliases: |
C11orf67, CK067, PTD015 |
Location: |
Chr.11:77821114-77901281 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 6 - Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AAMDC | NM_001316957.1 | NP_001303886.1 | ||
| NM_001316958.1 | NP_001303887.1 | |||
| NM_001316960.1 | NP_001303889.1 | |||
| NM_001316961.1 | NP_001303890.1 | |||
| NM_001316962.1 | NP_001303891.1 | |||
| NM_024684.3 | NP_078960.1 | |||
| NR_133637.1 | ||||
| XM_011544968.2 | XP_011543270.1 | |||
| XM_011544969.2 | XP_011543271.1 | |||
| XM_017017620.1 | XP_016873109.1 | |||
| AF092136.1 | AAD40378.1 | |||
| AI569731.1 | ||||
| AK024688.1 | BAB14963.1 | |||
| BC002752.2 | AAH02752.1 | |||
| BC016854.2 | AAH16854.1 | |||
| BI791786.1 | ||||
| BM930331.1 | ||||
| BP270361.1 | ||||
| BX646013.1 | ||||
| CN362204.1 | ||||
| CR457332.1 | CAG33613.1 |
Target Gene Details
Entrez Gene ID: | 51773 |
Gene Name: | remodeling and spacing factor 1 |
Gene Aliases: |
HBXAP, RSF-1, XAP8, p325 |
Location: |
Chr.11:77660009-77872232 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RSF1 | XM_017017923.1 | XP_016873412.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv520740 | Chr.11:77842592 - 78286890 on Build GRCh38 | Gain |
|
| nsv975953 | Chr.11:77867702 - 77870923 on Build GRCh38 | Duplication |
|
| nsv555453 | Chr.11:77866448 - 77907395 on Build GRCh38 | Loss |
|
| nsv1050891 | Chr.11:77705600 - 78076956 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map