Assay Details
Target Gene Details
Entrez Gene ID: | 27293 |
Gene Name: | sphingomyelin phosphodiesterase acid like 3B |
Gene Aliases: |
ASML3B |
Location: |
Chr.1:27934955-27959157 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SMPDL3B | NM_001009568.2 | 6 | 978 | NP_001009568.1 |
| NM_001304579.1 | 6 | 1049 | NP_001291508.1 | |
| NM_014474.3 | 6 | 978 | NP_055289.2 | |
| XM_011541259.2 | 7 | 1068 | XP_011539561.1 | |
| XM_011541260.2 | 7 | 1703 | XP_011539562.1 | |
| AK293489.1 | 5 | 680 | ||
| AK293566.1 | 6 | 975 | ||
| AK301349.1 | 6 | 933 | ||
| AK316500.1 | 6 | 904 | ||
| BC014444.1 | 6 | 895 | AAH14444.1 | |
| BX389470.1 | 2 | 111 | ||
| Y08134.1 | 6 | 870 | CAA69328.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs74467253] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map