Assay Details
Target Gene Details
Entrez Gene ID: | 55884 |
Gene Name: | WD repeat and SOCS box containing 2 |
Gene Aliases: |
SBA2 |
Location: |
Chr.12:118032687-118062430 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| WSB2 | NM_001278557.1 | 6 | 833 | NP_001265486.1 |
| NM_001278558.1 | 4 | 556 | NP_001265487.1 | |
| NM_018639.4 | 6 | 933 | NP_061109.1 | |
| XM_017019642.1 | 6 | 835 | XP_016875131.1 | |
| AF038187.1 | ||||
| AF163324.1 | 5 | 770 | AAF80478.1 | |
| AF229181.1 | 6 | 872 | AAF71302.1 | |
| AK294230.1 | 4 | 590 | ||
| AK295555.1 | 6 | 834 | ||
| AK298513.1 | 4 | 459 | ||
| AK310892.1 | 6 | 811 | ||
| BC015887.2 | 6 | 823 | AAH15887.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3581916 | Chr.12:117356518 - 118228514 on Build GRCh38 | Gain |
|
| nsv560388 | Chr.12:117953795 - 118044547 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map