Assay Details
Target Gene Details
Entrez Gene ID: | 56901 |
Gene Name: | NDUFA4, mitochondrial complex associated like 2 |
Gene Aliases: |
NUOMS |
Location: |
Chr.12:57234903-57240762 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NDUFA4L2 | NM_020142.3 | 4 | 862 | NP_064527.1 |
| XM_005269033.2 | 6 | 835 | XP_005269090.1 | |
| XM_011538573.2 | 5 | 1477 | XP_011536875.1 | |
| AF164796.1 | 4 | 607 | AAF80760.1 | |
| AK094275.1 | 8 | 1692 | ||
| AK129629.1 | 4 | 879 | ||
| BC011910.2 | 4 | 862 | AAH11910.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv482992 | Chr.12:56206217 - 57706217 on Build GRCh38 | Gain+Loss |
|
| nsv952189 | Chr.12:57127118 - 57245617 on Build GRCh38 | Deletion |
|
| nsv519293 | Chr.12:57189889 - 57243810 on Build GRCh38 | Loss |
|
| dgv96n21 | Chr.12:57139199 - 57243810 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map