Assay Details
Target Gene Details
Entrez Gene ID: | 9848 |
Gene Name: | microfibrillar associated protein 3 like |
Gene Aliases: |
NYD-sp9 |
Location: |
Chr.4:169986597-170033028 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MFAP3L | NM_001009554.3 | 2 | 795 | NP_001009554.1 |
| NM_001301647.1 | 2 | 870 | NP_001288576.1 | |
| NM_001301648.1 | 3 | 1057 | NP_001288577.1 | |
| NM_021647.7 | 3 | 1301 | NP_067679.6 | |
| XM_005263366.3 | 4 | 1235 | XP_005263423.1 | |
| XM_005263368.3 | 2 | 2409 | XP_005263425.1 | |
| XM_017008864.1 | 5 | 1179 | XP_016864353.1 | |
| XM_017008865.1 | 5 | 4295 | XP_016864354.1 | |
| XM_017008866.1 | 5 | 1369 | XP_016864355.1 | |
| XM_017008867.1 | 5 | 1175 | XP_016864356.1 | |
| XM_017008868.1 | 4 | 1450 | XP_016864357.1 | |
| XM_017008869.1 | 3 | 4282 | XP_016864358.1 | |
| XM_017008870.1 | 3 | 4242 | XP_016864359.1 | |
| XM_017008871.1 | 3 | 1182 | XP_016864360.1 | |
| AB014526.1 | 3 | 1149 | ||
| AF327560.1 | 2 | 781 | AAK15700.1 | |
| AK290041.1 | 3 | 1211 | ||
| AK308263.1 | 2 | 785 | ||
| AY391838.1 | 2 | 1082 | AAS55434.1 | |
| BC066912.1 | 2 | 764 | AAH66912.1 | |
| BM552945.1 | 2 | 807 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830149 | Chr.4:169811597 - 169999293 on Build GRCh38 | Loss |
|
| nsv1020188 | Chr.4:169856351 - 169992747 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map