Assay Details
Target Gene Details
Entrez Gene ID: | 83933 |
Gene Name: | histone deacetylase 10 |
Gene Aliases: |
HD10 |
Location: |
Chr.22:50245183-50251405 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 15 - Intron 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HDAC10 | NM_001159286.1 | NP_001152758.1 | ||
| NM_032019.5 | NP_114408.3 | |||
| AF393962.1 | AAK84023.1 | |||
| AF407272.1 | AAK92205.1 | |||
| AF407273.1 | AAK92206.1 | |||
| AF426160.1 | AAL30513.1 | |||
| AY450395.1 | AAS48345.1 | |||
| BC125082.1 | ||||
| BC125083.1 | ||||
| CR456465.1 | CAG30351.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv955199 | Chr.22:50149372 - 50320271 on Build GRCh38 | Deletion |
|
| nsv471223 | Chr.22:50190317 - 50271919 on Build GRCh38 | Loss |
|
| dgv781n67 | Chr.22:50153213 - 50314433 on Build GRCh38 | Gain |
|
| nsv1066496 | Chr.22:50226183 - 50292445 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map