Assay Details
Target Gene Details
Entrez Gene ID: | 283471 |
Gene Name: | transmembrane protease, serine 12 |
Gene Aliases: |
CT151 |
Location: |
Chr.12:50842918-50887884 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMPRSS12 | NM_182559.2 | 5 | 927 | NP_872365.1 |
| BC048112.1 | 5 | 912 | AAH48112.1 |
More Information
Set Membership: |
|

Genomic Map