Assay Details
Target Gene Details
Entrez Gene ID: | 200895 |
Gene Name: | dihydrofolate reductase 2 |
Gene Aliases: |
DHFRL1, DHFRP4 |
Location: |
Chr.3:94057922-94063656 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DHFR2 | NM_001195643.1 | 1 | 132 | NP_001182572.1 |
| XM_011512537.2 | XP_011510839.1 | |||
| AK122674.1 | 1 | 132 |
Target Gene Details
Entrez Gene ID: | 63899 |
Gene Name: | NOP2/Sun RNA methyltransferase family member 3 |
Gene Aliases: |
MST077, MSTP077 |
Location: |
Chr.3:94063011-94126786 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NSUN3 | NM_022072.3 | 1 | 82 | NP_071355.1 |
| AF169972.1 | 1 | 90 | AAQ13600.1 | |
| AK026262.1 | 1 | 74 | BAB15417.1 | |
| BC020602.1 | 1 | 82 | AAH20602.1 | |
| BC057238.1 | 1 | 86 | AAH57238.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3893746 | Chr.3:93792469 - 94164796 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map