Assay Details
Target Gene Details
Entrez Gene ID: | 122773 |
Gene Name: | kelch domain containing 1 |
Gene Aliases: |
MST025 |
Location: |
Chr.14:49693105-49753152 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KLHDC1 | NM_172193.2 | NP_751943.1 | ||
| XM_011536422.2 | XP_011534724.1 | |||
| AF111806.1 | AAL39008.1 | |||
| AK127202.1 | ||||
| BC101595.1 | AAI01596.1 | |||
| BC101597.1 | AAI01598.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1035708 | Chr.14:49632775 - 49749281 on Build GRCh38 | Gain |
|
| esv3634469 | Chr.14:49716658 - 49728339 on Build GRCh38 | Gain |
|
| esv3634466 | Chr.14:49643688 - 49728973 on Build GRCh38 | Gain |
|
| esv28926 | Chr.14:49426970 - 50475809 on Build GRCh38 | Gain+Loss |
|

Genomic Map